Specifications
- Reactivity:Human
- Description:Human HGF ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-576
Specifications
About this item
Human HGF AccuSignal ELISA Kit
- Natural and recombinant human HGF.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: sf21; Immunogen sequence: Q32-R494(alpha)&V495-S728(beta)
Hepatocyte growth factor(HGF) is the most potent mitogen for mature parenchymal hepatocytes in primary culture, and seems to be a hepatotrophic factor that acts as a trigger for liver regeneration after partial hepatectomy and liver injury. HGF has a relative molecular mass(Mr) of 82,000 and is a heterodimer composed of a large alpha-subunit of Mr 69,000 and a small beta-subunit of Mr 34,000. The protein consists of 728 amino acid residues, including a possible signal peptide at the N-terminus. HGF may serve as a paracrine mediator to control placental development and growth. This growth factor may play an important role as a paracrine mediator of the proliferation of melanocytes and endothelial cells, as well as cells of epithelial origin. The gene encoding the human HGF is assigned to human chromosome 7.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 8000pg/ml,4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml human HGF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(EDTA, citrate) to each empty well. It is recommended that each human HGF standard solution and each sample be measured in duplicate.