Specifications
- Reactivity:Human
- Description:Human Fractalkine - CX3CL1 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-616
Specifications
About this item
Human Fractalkine - CX3CL1 AccuSignal ELISA Kit
- Natural and recombinant human Fractalkine.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: G27-R189
Chemokine(C-X3-C motif) ligand 1(CX3CL1) is a large cytokine protein of 373 amino acids, it contains multiple domains and is the only known member of the CX3C chemokine family. It is also commonly known under the names fractalkine(in humans) and neurotactin(in mice). CX3CL1 is produced as a long protein(with 373-amino acid in humans) with an extended mucin-like stalk and a chemokine domain on top. The mucin-like stalk permits it to bind to the surface of certain cells. However a soluble(90 kD) version of this chemokine has also been observed. Soluble CX3CL1 potently chemoattracts T cells and monocytes, while the cell-bound chemokine promotes strong adhesion of leukocytes to activated endothelial cells, where it is primarily expressed. CX3CL1 elicits its adhesive and migratory functions by interacting with the chemokine receptor CX3CR1. Its gene is located on human chromosome 16 along with some CC chemokines known as CCL17 and CCL22. It can act as a mediator of smooth muscle cell migration in human atherosclerosis, rather than mediate inflammatory cell recruitment.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human Fractalkine standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA), saliva, urine or human milk to each empty well. It is recommended that each human Fractalkine standard solution and each sample be measured in duplicate.