Order Entry
ContactUsLinkComponent
Rat Fibronectin ELISA Kit, Rockland Immunochemicals
Rat Fibronectin ELISA Kit, Rockland Immunochemicals
Catalog #: 76236-608
CAS Number:  
Rat Fibronectin ELISA Kit, Rockland Immunochemicals
Catalog #: 76236-608
CAS Number:  

Specifications

  • Reactivity:
    Rat
  • Target protein:
    Fibronectin
  • Description:
    Rat Fibronectin ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76236-608

Specifications

About this item

Rat Fibronectin AccuSignal ELISA Kit

  • Natural rat Fibronectin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • From Plasma

Fibronectin(FN) also known as LETS, is identified on the surfFN of fibroblasts by labeling with radioactive compounds or specific antibodies. Fibronectin is a 430,000-dalton dimeric glycoprotein that exists in 2 forms, termed cellular and plasma fibronectin. Cellular and plasma fibronectins are heterodimers consisting of similar but not identical polypeptides. These two forms of FN differ in biologic activity. Fibronectins bind cell surfFNs and various compounds including collagen, fibrin, heparin, DNA, and actin. Because fibronectin stimulates endocytosis in several systems and promotes the clearance of particulate material from the circulation, it could function in the clearance of C1q-coated material such as immune complexes or cellular debris. Fibronectins are involved in cell adhesion, cell motility, opsonization, would healing, and maintenance of cell shape. LETS, encoded on chromosome 8, is responsible for the LETS protein expression in humans. Because LETS has been implicated in tumorigenicity and cellular transformation, it is of interest that rearrangement or modifications in the number of chromosome 8 have been associated with certain forms of cancer. The standard used in this kit is isolated from rat plasma with the molecular mass of 200-250KDa.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 313pg/ml, 156pg/ml rat Fibronectin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. We recommend that each rat Fibronectin standard solution and each sample is measured in duplicate.

Customers Who Bought This Also Bought