Specifications
- Reactivity:Human
- Description:Human sFAS ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76237-138
Specifications
About this item
Human sFAS AccuSignal ELISA Kit
- Natural and recombinant human sFAS.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: R17-N173
Fas, also known as APO-1, CD95 and TNFRSF6, is a member of the nerve growth factor(NGF)/tumour necrosis factor(TNF) receptor superfamily and mediates apoptosis. The nucleotide sequence of the cDNAs reveales that the molecule coding for the Fas antigen determinant is a 419 amino acid polypeptide with a single transmembrane domain. The extracellular domain is rich in cysteine residue, and shows a similarity to that of human tumor necrosis factor receptors, human nerve growth factor receptor, and human B cell antigen CD40. The APO-1 antigen as defined by the mouse monoclonal antibody anti-APO-1 is previously found to be expressed on the cell surface of activated human T and B lymphocytes and a variety of malignant human lymphoid cell lines. The APO-1 antigen is found to be a membrane glycoprotein of 48-kDa. Fas antigen is expressed and functional on papillary thyroid cancer cells and this may have potential therapeutic significance. Fas can play a role as an inducer of both neurite growth in vitro and accelerates recovery after nerve injury in vivo. The FAS and FASL triggered apoptosis pathway plays an important role in human carcinogenesis.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human FAS standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernatants, serum or plasma(heparin, EDTA, citrate) to each empty well. We recommend that each human FAS standard solution and each sample is measured in duplicate.