To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Freund's Complete Adjuvant
- Size:100 mL
- Cat. no.:CAD614-0100
- Supplier No.:D614-0100
Specifications
About this item
Freund's Complete Adjuvant (CFA)
- water-in-oil emulsions with antigens for the injection of immunogens for antibody production
- Autoclaved at 250°C
Special formulation of adjuvant containing paraffin oil, surfactant and heat-killed Mycobacterium butyricum.
Freund’s adjuvant may be used to form water-in-oil emulsions with antigens for the injection of immunogens for antibody production. Proper emulsion formation is essential and should be tested by observing a drop of the emulsion on the surface of water which should 'bead and not disperse. Antigens properly emulsified stimulate a strong and long-lasting immune response due to the slow release of antigen into the host’s system.