About this item
Rabbit Anti-GAPDH Antibody
- Affintiy Purified
- ELISA, Western Blot
GAPDH loading control antibody is ideal for Western Blotting, ELISA, IHC and IF Microscopy. GAPDH is constitutively expressed at high levels in almost all tissues and cell lines, making it ideal for use as a loading control antibody in immunoblots. A loading control antibody is critical for the correct interpretation of your western blot. Antibodies to loading controls are used to normalize the levels of protein detected by confirming that protein loading is uniform across the gel. Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) catalyzes the reversible oxidative phosphorylation of glyceraldehyde-3-phosphate in the presence of inorganic phosphate and nicotinamide adenine dinucleotide (NAD), an important energy-yielding step in carbohydrate metabolism. Recent evidence suggests that it also is involved in a number of cellular processes such as membrane fusion, phosphotransferase activity, DNA replication and repair, and nuclear RNA export. GAPDH has also been implicated in playing a role in different pathologies such as cancer progression, apoptosis, and neuronal diseases such as Alzheimer’s and Huntington’s disease. Anti-GAPDH antibody is ideal for investigators involved in apoptosis, cancer, DNA damage and repair and neuroscience.
Anti-GAPDH Antibody has been tested for use in ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at ~36 kDa in size corresponding to GAPDH by western blotting in the appropriate cell lysate or extract. GAPDH is constitutively expressed at high levels in almost all tissues and cell lines, making it ideal for use as a loading control marker. This Anti-GAPDH Antibody product is a new product replacing p/n 600-401-A33.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76262-718
- 76262-720
- Antigen symbol:
- GAPDH
- GAPDH
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Shipping temperature:
- Dry Ice
- Dry Ice