To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antigen symbol:IgG1
- Conjugation:Biotin
- Size:100 µg
- Shipping temperature:Ambient
- Cat. no.:76221-906
Specifications
About this item
Llama IgG1 isotype control Biotin Conjugate
- control or standard reagent
- ELISA, western blotting, Immunohistochemistry
Comparative studies of old world camelids (Camelus bactrianus and Camelus dromedarius) and new world camelids (Lama pacos, Lama glama and Lama vicugna) have shown that heavy-chain-only immunoglobulins represent between 35% - 70% of total IgG in the sera of all species. Such antibodies are homodimers of heavy chains that lack the CH1 domain of conventional antibodies and therefore do not interact with light chains, exhibiting a lower molecular weight ~100 kDa. In llama and other species of camelids, these heavy-chain-only immunoglobulins belong to the IgG2 and IgG3 subclasses.
All gamma chain camelid antibodies exhibiting the more conventional assembly of two light and two heavy chains with molecular weight ~150 kDa, belong to the IgG1 subclass.
Llama IgG1 Biotin Conjugated isotype control can be utilized as a control or standard reagent in Flow Cytometry, ELISA, and western blotting experiments where determination of sample isotype is important.