To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Gene ID:Q02750
- Protein/peptide type:Recombinant
- Source:Insect cells (Sf9)
- Species:Human
- Size:10 µg
- Storage conditions:Store vial at –20 °C prior to opening. This product is stable at 4 °C as an undiluted liquid. For extended storage, aliquot contents and freeze at –20 °C or below. Avoid cycles of freezing and thawing. Dilute only prior to immediate use.
- Protein synonyms:MEK1|MAP2K|MEK-1 recombinant protein|PRKMK1|MKK1|MEK
- Protein/peptide name:MAP2K1
- Purity:>90%
- Formulation:Liquid (sterile filtered)
- Shipping temperature:Dry Ice
- Cat. no.:76221-860
Specifications
About this item
MEK1 recombinant protein was produced in Sf9 cells. Mitogen-activated protein kinase kinase 1, also known as MKK or MEK, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. This pathway also plays a key role in synaptic plasticity in the brain. Activated MEK 1 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase. MEK1 and MEK2 are about 80% identical to each other, and nearly identical within the kinase domain. Recombinant MEK1 protein is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.