Avantor® Hichrom, Guard Cartridge Holder for HPLC Columns
Catalog #: 75830-798
Supplier: HICHROM LIMITED
Specifications
- Description:All-Guard Cartridge Holder for 2.1 - 4.6 mm I.Ø Columns
- Cat. no.:75830-798
- Supplier No.:80101/N
Specifications
About this item
This guard cartridge holder promotes increased column lifetime.
- Improved hardware
- Higher pressure rating
- Low internal swept volume
This guard cartridge holder is supplied in new hardware, leading to more robust and higher efficiency separations. Chromatographic selectivity will be unaffected by this change.
Customers Who Bought This Also Bought