Benzyl alcohol 98.0-100.5% (by GC), EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, SAFC®
Supplier: EMD MILLIPORE – PCS CA EXCEP
Certificates
About this item
Synonyms: Phenylmethanol
Formula:C₆H₅CH₂OH
MW:108.14 g/mol
Boiling Pt:205 °C (1013 hPa)
Melting Pt:-15.3 °C
Density:1.05 g/cm³ (20 °C)
MDL Number:MFCD00004599
CAS Number:100-51-6
:Storage Temperature: Ambient
ADR:9,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,0-100,5
Flash Pt:101 °C
:Boiling Pt:
:Boiling Pt: 205 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -15.3 °C
:Flash Pt:
:Flash Pt: 101 °C
:M.W. 108.14 g/mol
EINECS:202-859-9
:205 °C (1013 hPa)
:101 °C
:GHS07
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (GC):98.0 - 100.5%
- Identity (IR):Conforms
- Acidity:Conforms
- Refractive Index (n 20/D):1.538 - 1.541
- Density (d 20 °C/20 °C):1.043 - 1.049
- Peroxide Value:≤5.0
- Related Substances (GC) (Benzaldehyde (Impurity A)):≤0.15%
- Related Substances (GC) (Cyclohexylmethanol (Impurity B)):≤0.10%
- Related Substances (GC) (Sum of Peaks with a Relative Retention Less than that of Benzyl Alcohol):≤0.04%
- Related Substances (GC) (Sum of Peaks with a Relative Retention Greater than that of Benzyl Alcohol):≤0.3%
- Related Substances (GC) (Disregard Limit):≤0.0001%
- Benzene (GC):≤0.0002%
- Chlorobenzene (GC):≤0.01%
- Toluene (GC):≤0.01%
- Other Residual Solvents (ICH Q3C):Excluded by Manufacturing Process
- Evaporation Residue:≤0.050%
- Water:≤0.10%
Specification Test Results
Customers Who Bought This Also Bought