Digital Hotplates, SuperNuova+™, Barnstead, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
These SuperNuova+™ hotplate series provides reliable performance, dependable safety and simple operation.
- Top plate choices in ceramic or aluminum
- Easy-to-read LED display for heating
- Raised display design protects electronics from spills
- Ability to save temperature setting
These hotplate units are available in two sizes, including a multi-position model with optimized controls and settings for applications that demand advanced precision.
The internally designed hot top warning system protects from accidental burns with prominent display when heating surface is above 50 °C (122 °F).
Certifications: cCSAus, CE compliance.
Delivery: Supplied with PT100 temperature probe, rod and clamps. Optional PT1000 temperature probe and splash guard protection shield needs to be ordered separately.
Customers Who Bought This Also Bought