CELLdisc™ MultiLayer Cell Culture Systems, Greiner Bio-one
Supplier: Greiner Bio-One
Certificates
About this item
CELLdisc™ is a multilayer device covering a range of cell culture surfaces from 250 cm2 up to 10000 cm2. The innovative, ergonomic design provides a versatile system from research scale to industrial batches.
- Non-pyrogenic, non-cytotoxic
- Free from DNase, RNase, and human DNA
- Screw cap color indicating surface treatment
- Integrated gas support channel and vent port for pressure equilibration
- Available in 4-, 8-, 16-, and 40-layer versions
A centrally located channel allows uniform distribution of gas throughout the device. A socle-rim guarantees that the bottom layer of the CELLdisc™ does not touch the surface of the incubator. These compact and robust cylindrical devices are ideally suited for automation and upscaling of mass cell culture. A large 38mm opening with screw cap enables easy filling.
Certifications: USP Class VI and Sterility Assurance Level (SAL) of 10–6.
Related Information
Certificates
Search by Lot Number75870-806_E20023H6
Conformity declaration
Greiner Bio-One Product Catalog
Product Literature
Customers Who Bought This Also Bought