Lattice Lab System Set #1, United Scientific Supplies
Catalog #: 76293-332
Supplier: United Scientific Supplies
Specifications
- Description:Lattice Lab System Set #1
- Cat. no.:76293-332
Specifications
About this item
The Lattice Lab System is a lightweight set of aluminum support rods and clamps that allows a wide range of lab apparatus to be assembled and securely supported.
- Sturdy aluminum construction
- Includes mounting hardware to tables
Assembly is easy and requires only a screwdriver. Support rods are made of hard aluminum and foot plates and hook connectors are made of cast alloy.
Delivery: Set #1 includes: (4) Foot Plates, 2.3125" diameter.; (4) Set Screws for Foot Plate; (12) Anchoring Screws for Foot Plate; (22) Hook Connectors with Thumbscrew; (8) Support Rods, 2" L, 0.5" diameter.; (2) Support Rods, 18" L, 0.5" diameter.; (8) Support Rods, 24" L, 0.5" diameter.
Customers Who Bought This Also Bought