Crucible Tongs, Plated Steel, United Scientific Supplies
Catalog #: 76293-542
Supplier: United Scientific Supplies
Specifications
- Length:9"
- Description:Crucible Tongs
- Material:Plated Steel
- Cat. no.:76293-542
Specifications
About this item
Tongs have an overall length of 9" and include riveted joints and serrated tips.
- Overall length is 9"
- Riveted joints and serrated tips
Pick up hot crucibles safely. The overall 9" length keeps you at a safe distance ion moving hot crucibles, all while maintaining a controlling grip.
Customers Who Bought This Also Bought