Mouse Recombinant SCF (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
Customers Who Bought This Also Bought
Sodium hydroxide 0.02 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
73,05 $ to 828,17 $
Sodium hydroxide 50% (w/w) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
220,41 $ to 875,50 $
Fendall® Saline Eyewash Wall Stations, Honeywell Safety
Supplier: Honeywell Safety Products
226,73 $ to 1 769,47 $
HyFlex® 11-318 Touch Screen-Capable Gloves, Uncoated, Ansell
Supplier: Ansell Healthcare
300,30 $ to 3 498,74 $
uTIP™ CleanPak™ Pipette Tip Reload System for Universal Fit Pipettes, Biotix Inc
Supplier: Biotix
81,72 $ to 326,77 $
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
500,19 $ to 1 790,36 $