Specifications
- Dimensions:6×9"
- Color:Grey
- Description:Hand pad 7448, ultra fine
- Cat. no.:21908-072
- Supplier No.:7448
Specifications
About this item
Wide array of hand pads for cleaning, scuffing, deburring, finishing and other surface conditioning applications.
- Tightly graded with more active mineral sites for fast, consistent action without compromising your final finish
- Flexible construction is conformable for more complete scratch in channels, grooves, tight spots and other hard-to-reach areas
- Durable, specially-coated to resist loading and prolong pad life
Scotch-Brite™ Hand Pads give you a fast, consistent scuff for everyday surface preparation. Ideal for cleaning parts and Hand pads are also a great alternative to steel wool — similar performance without the rusting or splintering.
7447: Worldwide standard for surface conditioning; Clean, blend, finish and deburr faster, easier, and with better results than conventional abrasives. Performance similar to steel wool grade 1.
7445: Very mild abrasive for gentle, yet thorough cleaning. Commonly used with liquid detergent. Performance similar to steel wool grade 0000.
7440: Most durable and aggressive pad for quick stock removal. Cuts fast to remove contaminants and oxides in demanding cleaning and finishing applications. Performance similar to steel wool grade 4.
7448: Ultra fine finishing action outperforms steel wool for cleaning, blending, prepping and finishing on metal, wood, plastics and composites. Performs like steel wool grade 00 without shredding, rusting, or splintering.
96-20: Medium duty scouring pad made from high quality fibers, mineral, and resin. Designed to be used for everyday cleaning of most cooking utensils and equipment.