ADC® Adscope® 600 Platinum Cardiology Stethoscopes
Catalog #: 470309-764
Supplier: AMERICAN DIAGNOSTIC CORP CP
Specifications
- Color:Black
- Description:Adscope 600
- Cat. no.:470309-764
Specifications
About this item
The best acoustic response available!
- Assembled, inspected, and packaged in the USA
- Lifetime warranty covers all parts
- Accessory kit includes two spare pairs of Adsoft™ Plus ear tips
The ADC® Adscope® 600 is engineered for performance, comfort, and durability. The bi-lumen (dual channel) headset optimizes the acoustic pathway while the adjustable frequency (AFD) technology, housed in a compact, sculpted design, allows you to enhance both high- and low-frequency responses. The ergonomic cardiology headset, made with heavy-walled tubing, large-diameter aural tubes, and a double-leaf tension spring, wears easily while withstanding thousands of cycles. The Cardiology Series features an all stainless steel construction, ensuring a lifetime of use.
Customers Who Bought This Also Bought