Sodium dihydrogen phosphate monohydrate 98.0-103.0% (by anhydrous basis) USP, ACS, J.T.Baker®
Catalog #: CAJT3808-65
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 10049-21-5
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:HDPE drum, Flowmor™ package
- Size:100 kg
- Cat. no.:CAJT3808-65
Specifications
Specification Test Results
- GMP Manufactured Product:
- Meets ACS Reagent Chemical Requirements:
- Meets U.S.P Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Assay (anhydrous basis):98.0 - 103.0 %
- USP - Aluminum, Calcium, and Related Elements:Passes Test
- USP - Arsenic (As):≤8 ppm
- USP - Chloride (Cl):≤0.014 %
- USP - Identification:Passes Test
- USP - Insoluble Substances:≤0.2 %
- USP - pH (1 in 20):4.1 - 4.5
- USP - Sulfate (SO₄):≤0.15 %
- USP - Water (H₂O):10.0 - 15.0 %
- Assay (NaH₂PO₄·H₂O):98.0 - 102.0 %
- pH of 5% Solution at 25°C:4.1 - 4.5
- Insoluble Matter:≤0.01 %
- Chloride (Cl):≤5 ppm
- ACS - Sulfate (SO₄):≤0.003 %
- Calcium (Ca):≤0.005 %
- Potassium (K):≤0.01 %
- Heavy Metals (as Pb):≤0.001 %
- Trace Impurities - Iron (Fe):≤0.001 %
- For EP notated requirements, results are derived from testing Sodium:
- Phosphate, Monobasic Monohydrate, U.S.P. to the EP Chemical:
- specifications for Sodium Dihydrogen Phosphate Dihydrate.:
- For JPE notated requirements, results are derived from testing Sodium:
- Phosphate, Monobasic Monohydrate, U.S.P. to the JPE Chemical:
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- J.T.Baker®, Sodium dihydrogen phosphate monohydrate 98.0-103.0% (by anhydrous basis) USP, ACS:
Specification Test Results
About this item
Synonyms: Sodium phosphate monobasic monohydrate,Sodium dihydrogen orthophosphate monohydrate
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,0-103,0
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 100 °C
:Flash Pt:
:Flash Pt:
:M.W. 137.99 g/mol
:100
:12,08806
Formula:NaH₂PO₄·H₂O
MW:137.99 g/mol
MDL Number:MFCD00149208
CAS Number:10049-21-5
Customers Who Bought This Also Bought