ContactUsLinkComponent
Ward's® Chromatography of Amino Acids
Ward's® Chromatography of Amino Acids
Catalog #: 470225-450
Supplier:  Ward's Science
Chromatography Of Amino Acids
Ward's® Chromatography of Amino Acids
Catalog #: 470225-450
Supplier:  Ward's Science
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Chromatography of Amino Acids
  • No. of Lab Groups:
    15
  • Cat. no.:
    470225-450
  • Supplier No.:
    AD-KK7108-KIT
Specifications
About this item
Do you know how to separate proteins?
  • Middle or high school activity
  • Aligned to NGSS Standards PS1.A and PS1.B
  • Contains a teacher's guide and study study guide copymasters
Amino acids are the fundamental building blocks of all proteins. Often times scientists may need to identify the amino acid contained in a protein. One such method for separating and identifying individual amino acids is paper chromatography.
In this activity, students will perform a paper chromatography experiment on three known amino acids, and then use their results to identify the components of an unknown amino acid mixture.
Caution: This product can expose you to chemicals including Methanol/Methyl isobutyl ketone, which is known to the State of California to cause cancer, birth defects and reproductive harm. For more information go to http://www.P65Warnings.ca.gov.
Customers Who Bought This Also Bought