Industrial Evaporator Flasks, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Large receiver/evaporator flasks are designed for use with Heidolph 20L rotary evaporators.
- Flasks are fabricated from blanks selected for balance and quality, with carefully welded necks to prevent "rotational whip"
- Flask has a polished 40/25 joint, and side neck is a GL-18 thread with a solid cap
- Flask bottom has a 0–10mm easy action stopcock with a GL-18 arm which comes supplied with a 3/8" connection tube
Customers Who Bought This Also Bought