tri-Sodium citrate dihydrate 99.0-100.5% (by anhydrous basis), crystalline, GenAR® USP, Macron Fine Chemicals™
Catalog #: CAMK063445
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 6132-04-3
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Fibre drum
- Size:125 kg
- Cat. no.:CAMK063445
Specifications
Specification Test Results
- GMP Manufactured Product:
- Meets E.P. Chemical Specifications:
- Meets U.S.P Requirements:
- Meets J.P. Chemical Specifications:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Assay (C₆H₅Na₃O₇) (anhydrous basis):99.0 - 100.5 %
- USP - Appearance (colorless or white to slightly yellow crystals):Passes Test
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Identification C:Passes Test
- USP - Alkalinity:Passes Test
- USP - Water (H₂O):10.0 - 13.0 %
- USP - Tartrate:Passes Test
- EP - Assay (anhydrous basis):99.0 - 101.0 %
- EP - Appearance of Solution (clear and colorless):Passes Test
- EP - Identification A:Passes Test
- EP - Identification B:Passes Test
- EP - Acidity or Alkalinity:Passes Test
- EP - Readily Carbonizable Substances:Passes Test
- EP - Chlorides (Cl):≤50 ppm
- EP - Oxalate (C₂O₄):≤300 ppm
- EP - Sulfate (SO₄):≤150 ppm
- EP - Water (H₂O):11.0 - 13.0 %
- JP - Assay (anhydrous basis):99.0 - 101.0 %
- JP - Description:Passes Test
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - pH:7.5 - 8.5
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤0.015 %
- JP - Sulfate (SO₄):≤0.048 %
- JP - Heavy Metals:≤10 ppm
- JP - Arsenic (As):≤2 ppm
- JP - Tartrate:Passes Test
- JP - Oxalate (C₂O₄):Passes Test
- JP - Readily Carbonizable Substances:Passes Test
- JP - Loss on Drying at 180°C:10.0 - 13.0 %
- Not Intended for Parenteral Use:
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- Macron Fine Chemicals™, tri-Sodium citrate dihydrate 99.0-100.5% (by anhydrous basis), crystalline, GenAR® USP:
Specification Test Results
About this item
Synonyms: Sodium Citrate Dihydrate,Sodium citrate tribasic dihydrate,Sodium Citrate, Dihydrate
:Storage Temperature: Ambient
Melting Pt:150 °C
Density:1.70 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 150 °C
:Flash Pt:
:Flash Pt:
:M.W. 294.1 g/mol
:tri-Sodium citrate dihydrate
Formula:Na₃C₆H₅O₇·2H₂O
MW:294.1 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00150031
CAS Number:6132-04-3
EINECS:200-675-3
:M.O.C. Category R
Customers Who Bought This Also Bought