Jacketed Beakers/Baths, Low Form, Flat Bottom, Chemglass
Catalog #: 89028-334
Supplier: Chemglass
Supplier #: CG-1107-11
Small Business Enterprise
Log in for availability
About this item
Jacketed beakers/baths with a larger opening at the top of the jacket and a shorter overall height.
- Width to Height ratio on all sizes is 2:1
- Flat bottom on both the inner and outer vessel
- Designed for use in temperature controlled chemical and biological reactions
Hose connections have an O.D. of 10 mm at the largest serration.
Customers Who Bought This Also Bought