Drechel Gas Washing Bottles, Tall Form, Chemglass
Supplier: Chemglass
Certificates
About this item
Tall form gas washing bottle having a standard taper 34/28 top joint, hex base, and plain tube for gas dispersion.
Stopper is held in place with two 18 mm stainless steel springs.
Note: springs are only supplied when bottle is ordered complete.
Customers Who Bought This Also Bought