ContactUsLinkComponent
First Aid Only Butterfly Bandages, Acme United
First Aid Only Butterfly Bandages, Acme United
Catalog #: 76287-288
Supplier:  First Aid Only
Butterfly Bandage Secure Wound 100/ BOX
First Aid Only Butterfly Bandages, Acme United
Catalog #: 76287-288
Supplier:  First Aid Only
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Butterfly Bandages
  • Cat. no.:
    76287-288
Specifications
About this item
Butterfly wound closures are latex-free and eliminate the need for stitches.
  • Close and secure small wounds
Butterfly wound closures are designed to close and secure small wounds and incisions while holding the skin firmly. They will not stick to the wound itself. These bandages remove the need for stitches in relatively minor cuts and they are sterile unless package is opened or damaged. Promotes fast healing as well and ensures a secure closure.
Packaging: Packed 100 per box.
Customers Who Bought This Also Bought