To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Total RNA
- Sample type:Plant samplesBacteriaBloodYeast/FungiCultured cellsHuman/animal cells and tissues
- Target molecule:RNA
- Description:Monarch® Total RNA Miniprep Kit
- Size:50 preps
- Storage conditions:25 °C
- Description:Monarch® Total RNA Miniprep Kit
- Cat. no.:CA103529-148
Specifications
About this item
Quickly and easily purify up to 100 µg of high quality total RNA from multiple sample types – all with one kit.
- Activity is not inhibited by an α1-3 Fucose residue on the chitobiose core
- Effectively purifies total RNA of all sizes, including small RNAs >20 nt
- Efficient genomic DNA removal (column and DNase I-based)
- Excellent value
The Monarch Total RNA Miniprep Kit is a comprehensive solution for sample preservation, cell lysis, gDNA removal, and purification of total RNA from a wide variety of biological samples, including cultured cells, blood, and mammalian tissues.
Additionally, tough-to-lyse samples, such as bacteria, yeast, and plant, can be processed with additional steps that enhance lysis. Cleanup of enzymatic reactions or purification of RNA from TRIzol® -extracted samples is also possible using this kit.
Purified RNA has high quality metrics, including A260/280 and A260/230 ratios >1.8, high RIN scores, and minimal residual gDNA. Captured RNA ranges in size from full-length rRNAs down to intact miRNAs. Additionally, differential binding conditions allow selective capture or exclusion of the sub-200 nucleotide RNA pool that includes miRNA, 5S rRNA, and tRNA. Purified RNA is suitable for downstream applications, such as RT-qPCR, cDNA synthesis, RNA-seq, Northern blot analysis.