Full UV/VIS Photometric Accuracy, Wavelength and Stray Light Calibration Kit, Fireflysci
Catalog #: 76016-850
Supplier: FIREFLYSCI, INC.
CAS Number:
Specifications
- Description:Full UV/VIS Photometric Accuracy, Wavelength & Stray Light Calibration Kit 4
- Cat. no.:76016-850
- Dimensions:12.5×12.5 mm
Specifications
About this item
Broad range solid-state standard kit for checking photometric accuracy, wavelength accuracy and stray light in the VIS range.
- Tolerances exceeding NIST 930e standards, pinpoint accuracy is assured
- No need to recalibrate due to material aging
- Resilient scratch-resistant coating ensures life-time longevity
- For use with all type of research-grade UV/VIS/NIR spectrophotometers
This calibration kit can test the following:
Photometric Accuracy in the UV and VIS range (HF-0.2, HF-0.5, HF-1.0, FUV-0.5) - wavelength points of 250, 280, 340, 360, 400, 465, 500, 546.1, 590 and 635nm included on the certificate.
Wavelength Accuracy in the UV and VIS range (WAV-1 UV/VIS, WAV-7 UV/VIS) - Data points of 241 279, 287, 329 333. 360, 385, 418, 453, 459, 472, 512, 536 and 637, 681, and 875nm included on the certificate.
Stray Light Performance in the VIS range (FUV-0.5).
Certifications: NIST compatible.
Delivery: The instructions manual is included.
Customers Who Bought This Also Bought