Ozone Vacu-vials Instrumental Kit, CHEMetrics
Catalog #: CXK-7423
Supplier: CHEMetrics
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Chemical method:DPD (N,N-diethyl-p-phenylenediamine)
- Description:Ozone Vacu-vials Instrumental Kit
- Range:0 - 5.00 ppm
- Supplier No.:K-7423
- No. of Tests:30
Specifications
About this item
This kit requires the use of the V-2000 or V-3000 Photometer, a SAM Photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell.
- Analysis time: 1 minute
- DPD method, 0 to 5.00 ppm
Delivery: It contains everything needed to perform 30 tests: thirty ampoules, activator Solution, 25 ml sample cup, ampoule blank, and instructions.
Packaging: Ozone Test Kit comes in a cardboard box.
Customers Who Bought This Also Bought