DuPont™ ProClean® 2 Heavy Duty Shoe Covers with Traction
Supplier: Avantor
Certificates
About this item
DuPont™ ProClean® 2 shoe covers are made from spunbond meltblown spunbond (SMS) with a traction sole.
- SMS shoe covers
- Serged seams
- Elastic opening
- Heavy duty material
Shoe covers feature a traction sole with serged seams and an elastic opening. Multiple packaging options are available.
Certifications: Thickness meets ASTM D3776.
Packaging: Shoe covers in Multi Pack option are packaged in a multi-pack dispenser with 400 per case. Shoe covers in Bulk Packed option are bulk packaged and placed in transparent poly liners with 500 covers per case.
Customers Who Bought This Also Bought