Specifications
- Description:Hydrion Lo Iodine Test Kit
- Cat. no.:89370-912
- Supplier No.:IL-250
Specifications
About this item
The Hydrion Lo iodine kit provides an easy way to estimate the amount of titratable iodine in sanitizing solutions made with commercial iodophors. The color chart is easy to read and has color matches at 0, 12.5, 25, 50 ppm (parts per million) of titratable iodine.