BBL™ DrySlide™ Nitrocefin, BD
Catalog #: CA-90002-576
Supplier: BD
Specifications
- No. of Slides:25
- Description:BBL™ DrySlide™ Nitrocefin
- Supplier No.:231749
Specifications
About this item
A disposable slide containing a reagent cassette impregnated with nitrocefin for the rapid qualitative detection of beta-lactamase producing bacteria.
- Test results in 60 minutes or less
Customers Who Bought This Also Bought