To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Powdered Yeast Media, pH ranges 4.4 ± 0.8
- Size:227 g (0.5 lbs.)
- Storage temperature:15…30 °C (59…86 °F)
- Cat. no.:76203-084
Specifications
About this item
YNB is a well defined composition of salts and vitamins for a vigorous growth of S. cerevisiae.
- Soluble in water (1.7 g/l)
Reagents are tested and chosen for their ability to promote optimum growth of S. cerevisiae when combined in YNB formulations. Chosen reagents are combined in the proper proportions according to the formulation listed above and thoroughly blended for a uniform distribution.
Add 1.7 g/L YNB without ammonium sulfate, plus a nitrogen source and a carbon source, the approproate amino acids, and agar if necessary to make your own SD yeast media.
Application includes: Yeast Cell Culture, Gene Expression, Transformation, Gene Function, Gene Regulation.