To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Bacterial Media, Acceptable pH range is 6.7 ±0.3 at 35 °C
- Size:454 g (1 lbs.)
- Storage temperature:Store sterile LB Agar, Lennox at ambient temperature (15 - 30 °C). Powder medium is hygroscopic, therefore minimize exposure to air to prolong shelf life of media.
- Cat. no.:CA76046-354
Specifications
About this item
Rich medium for the growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium.
- Proprietary media for enhanced cell yield
- Convenient packaging formats for a variety of applications and budgets
- Proven and reliable reputation
- High-quality raw materials and additives
- Encapsulated media
- No weighing, dust and cleanup
- Drop capsules into water and autoclave
LB lennox agar capsules is a nutritionally rich medium originally developed for growth and maintenance of E. coli strains. The ultimate in ease and convenience, LB Lennox Agar in a capsule form eliminates the waste, inaccuracies, and mess associated with weighing out bulk powder. The growth characteristics are the same as LB Lennox Agar powder. LB lennox agar medium supports a luxurious growth of E. coli cells as tryptone and yeast extract supply essential growth factors that E. coli cells would otherwise have to synthesize. The medium also contains essential electrolytes for transport and osmotic balance, due to the NaCl component.
Application includes: Bacteria cell culture, gene expression, transformation, microbiological analysis, protein expression.