To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Format:Caps
- Description:Lab Essential
- Size:5000 each
- Cat. no.:76203-086
Specifications
About this item
Green caps fit EMPTY FASTPREP® tubes and are used in fastprep systems, to hold sample for homogenization.
- Lysing Matrix D tubes have green caps found in FastRNA™ Pro green kit which are used for isolation of total RNA from plants and animals
- DNase and RNase-free
- Store at room temperature(15 - 30 °C)
Use with Empty FastPrep Tubes. Add sample to empty tube, add appropriate Lysing Matrix material and lysing buffer, and apply cap. Process using FastPrep instrument or other bead-beater to lsye, grind or homogenize sample.
Application includes: Sample preparation, sample lysis, nucleic acid isolation and purification, protein isolation, molecular biology, gene expression, microbiological analysis, gene function, gene regulation.