SpectroBlend® additive, tablets, Chemplex® 44 µm
Catalog #: 15566-083
Supplier: Chemplex
CAS Number:
Specifications
- Packaging:500/Bottle
- Size:0.5 g (500 Tablets/ Bottle)
- Cat. no.:15566-083
- Supplier No.:690
Specifications
Specification Test Results
- SpectroBlend® additive, tablets, Chemplex® 44 µm:
Specification Test Results
About this item
:Length:
:Width:
:Height:
:Ø
:Thickness:
:44 µm
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
Customers Who Bought This Also Bought