Magid® TouchMaster® 661H Medium Weight Hemmed Lisle Gloves, Magid
Catalog #: 32889-458
Supplier: Magid Glove
Small Business Enterprise
Specifications
- Ambidextrous/Hand-specific:Ambidextrous
- Glove size:M
- Powdered/Powder-free:Powder-free
- Description:Magid® TouchMaster® Medium Weight Hemmed Lisle Gloves
- Latex/Latex-free:Latex-free
- Cat. no.:32889-458
- Supplier No.:660H
Specifications
About this item
Medium weight cotton lisle inspection gloves are 100% cotton two-piece lisle inspection glove constructed of cool and breathable bleached white cotton.
- Color-coded sizes for easy sorting
- Hemmed edge to prevent unraveling
- Cut and sewn for the most comfortable fit
- General-purpose glove for light jobs in any industry
This glove can be worn alone to protect delicate products from smudges, finger prints or skin oils, or it can be used as a liner to absorb moisture.
Customers Who Bought This Also Bought