Uvex® V-Maxx Wrap-Around Goggles with Pivoting Headstrap, Magid Glove
Catalog #: 69100-546
Supplier: Magid Glove
Small Business Enterprise
Specifications
- Lens Coating:Anti-fog
- Description:Standard glass with elastic strap, direct vent
- Frame Color:Clear
- Cat. no.:69100-546
- Supplier No.:WVMAXXCI
Specifications
About this item
Uvex V-Maxx Safety Goggle offers superior eye protection and attractive futuristic styling. With its sleek wrap-around styling and 180°, clear field of vision, this offers top of the line protection with great visibility.
- Adjustable elastic or neoprene headband
- Pivoting headband clips to adjust strap around hard hats or hearing protection
Safety goggles feature direct and indirect ventilation styles available for impact or splash applications.
Certifications: Meets ANSI Z87+ and CSA Z94.3 safety standards.
Customers Who Bought This Also Bought