Magid® Kevlar® M93 Nitrile Dotted Gloves, Magid
Supplier: Magid Glove
Certificates
About this item
The Magid N93 is a seamless terrycloth glove featuring nitrile dots on one side of the glove.
- Reversible style means double the wear
- Single side covered with dots to ensure added wear and superior grip
- Overcast rubber band wrist prevents unraveling, keeps tight fit after washing
This durable style features a heavyweight loops-in terry knit, meaning the same cut and abrasion resistance with a reduced chance of snags.
Specifications
- Catalog No:
- 69100-512
- Ambidextrous/Hand-specific:
- Hand-specific
- Disposable / reusable:
- Reusable
- Latex/Latex-free:
- Latex-free
- Powdered/Powder-free:
- Powder-free