Magid® Softside Direct Vent Goggle, Magid Glove
Catalog #: 66200-106
Supplier: Magid Glove
Small Business Enterprise
Specifications
- Lens Coating:Scratch-resistant/hardcoat
- Lens Tint:Clear
- Description:Softside direct vent goggle
- Frame Color:Blue tint
- Supplier No.:250
Specifications
About this item
Magid® direct vent safety goggles feature with clear lenses and direct ventilated frames for splash resistance.
- Comfortable soft-side goggle
This provides exceptional value with a comfortable fit and allow for good peripheral vision.
Certifications: This high impact safety glasses meet ANSI Z87 high impact standards.
Customers Who Bought This Also Bought