Magid® CleanMaster® Lightweight Nylon Tricot Gloves, Magid
Supplier: Magid Glove
Certificates
About this item
The Magid CleanMaster is used for cleanroom, paint and electronics settings.
- Light weight two piece pattern
- Reversibility allows for glove to fit either hand
- Perfect for general-purpose use
This useful glove is designed for low lint, causing less product contamination.
Specifications
- Catalog No:
- 32933-916
- 32933-914
- 32933-918
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Material:
- Nylon
- Nylon
- Nylon