Ward's Cool Containers
Supplier: Heathrow Scientific
Certificates
About this item
Durable containers for chilling temperature-sensitive samples.
- Ideal for ice and ice/salt slurries
- Lid nests under bucket for convenience and to save space
- Tapered design for easy stacking
- Designed for low temperature use to –196 °C at 1 bar
Ward's cool containers are perfect for the laboratory or classroom. They are made from polyurethane with metal inserts for stability. The close-fit lid keeps evaporation to a minimum and features a comfortable recessed knob for easy handling. The precise pour spout and large handles allow for excess liquid to be removed easily.
Customers Who Bought This Also Bought