Specifications
- No. of Slides:15 slides - 15 ampules
- Description:PYR Kit
- Cat. no.:CA-90002-572
- Supplier No.:231747
Specifications
About this item
DrySlide™ PYR is a disposable slide format for the presumptive identification of group A streptococci and enterococci.
- Easy-to-handle slide format
- Each slide has four large segments, allowing multiple testing on one slide
Contains the reagent L-pyrrolidonyl-ß-naphthylamide (PYR) which is hydrolyzed by bacteria possessing the enzyme pyrrolidonyl amino peptidase. The addition of the DrySlide™ PYR color developer results in a vivid color reaction in a three minute test.