Medium Length Sleeves, PTFE, Chemglass
Supplier: Chemglass
Certificates
Certificates
About this item
These sleeves are made of PTFE for standard taper joints.
- Reusable
- Provides grease-free seal
Thickness: 0.005"
Customers Who Bought This Also Bought
Bytac® Teflon® Resin Surface Protectors, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
$1,747.88
SP Bel-Art Covamat™ Clear Bench Liner, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$362.33
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
$271.77
Nalgene® Polyolefin Plastic Paper Sheets, Thermo Scientific
Supplier: Thermo Fisher Scientific
$534.61 to $1,523.31