FLIPR Tetra Pipette Tips, Molecular Devices
Supplier: MOLECULAR DEVICES LLC. CA
Certificates
About this item
A comprehensive line of tips for Molecular Devices FLIPR Tetra automation systems.
- Ensure assay quality with quality tips
- Certified DNase-/RNase-free
- Non-pyrogenic
- Non sterile
Official pipette tips for the FLIPR Tetra High-Throughput Cellular Screening System. Tips are manufactured specifically to work under strict controls to guarantee optimal preformance and quality.
Customers Who Bought This Also Bought