Deoxynucleotide Mix, Agilent Technologies
Catalog #: CAST200415
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Deoxynucleotide Mix, PCR-Grade
- Size:0.4 ml
- Cat. no.:CAST200415
Specifications
About this item
The PCR grade deoxynucleotide mix includes 25 mM of each standard dNTP (GATC) for use with any PCR application, including real-time quantitative PCR.
- Ideal for optimal PCR performance
- Produces high-quality reaction products
- Withstands multiple freeze-thaw without compromising efficiency
Delivery: Includes 400 µl of 100 mM dNTP mix (25 mM of each dNTP), adequate for 1000 standard 50 µl primer-extension reactions or 2000 reactions of 25 µl.
Customers Who Bought This Also Bought