Order Entry
ContactUsLinkComponent
Multi-Neurotrophin Rapid Screening ELISA Kit, Human, Biosensis®
Multi-Neurotrophin Rapid Screening ELISA Kit, Human, Biosensis®
Catalog #: 75838-852
Supplier:  Biosensis
CAS Number:  
Multi-Neurotrophin Rapid Screening ELISA Kit
Multi-Neurotrophin Rapid Screening ELISA Kit, Human, Biosensis®
Catalog #: 75838-852
Supplier:  Biosensis
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugate ELISA:
    HRP
  • Reactivity:
    Human
  • Sensitivity:
    Typical LODs achieved based on 150% of background OD: NGF: 3 pg/ml BDNF: 1 pg/ml NT3: 4 pg/ml NT4/5: 10 pg/ml
  • Target protein:
    Neurotrophins: BDNF, NGF, NT3, NT4/5
  • Description:
    Multi-Neurotrophin Rapid Screening ELISA Kit: Human
  • Size:
    48 Tests
  • Sample type:
    Cell culture supernatants, lysates and brain extracts
  • Cross reactivity:
    This Multi-Neurotrophin Screening kit uses the same antibodies as in the individual Rapid ELISA kits. Each kit has been tested for cross-reactivity with other closely related neurotrophins and no cross-reactivity has been observed at 25 ng/ml of each neurotrophin tested.The antibodies used in this kit detect the mouse and rat form of each neurotrophin, but with varying efficiency depending on the target protein
  • Detection method:
    Photometric (450 nm)
  • Specificity:
    Each kit has been tested for cross-reactivity with other closely related neurotrophins and no cross-reactivity has been observed at 25 ng/ml of each neurotrophin tested
  • Detection range:
    NGF (3.9 - 250 pg/ml) BDNF (7.8 - 500 pg/ml) NT3 (15.6 - 1000 pg/ml) NT4/5 (15.6 - 1000 pg/ml)
  • Cat. no.:
    75838-852
  • Supplier No.:
    BEK-2227

Specifications

About this item

The biosensis® Multi-Neurotrophin RapidTM Screening ELISA kit has been designed to allow rapid screening and quantification of human NGF, BDNF, NT3 and NT4/5 in cell culture supernatants, lysates and brain extracts only if used as directed. Caution should be exercised for assaying neurotrophins in blood.

Please refer to the kit protocol for specific use instructions for each substrate application, in particular human serum and plasma samples. This two-plate kit consists of four sets of 6 strips for each Neurotrophin, allowing for the assay of 16 samples per Neurotrophin tested and a full range of standards, all run in duplicate. It provides the identical sensitivities and ranges that are achieved in the complete, individual ELISA kits, thus allowing easy progression into the larger individual assay sizes when needed.

The Multi-Neurotrophin RapidTM Screening ELISA kit therefore presents a cost-effective way to quickly screen multiple samples, which can then be published or used prior to a more extensive analysis with the individual kits. The kit comes complete with all detection reagents and is ready to use. Individual coated strips are provided, and each set of standards and detection antibodies are color-coded.

Delivery: The ELISA kit box contains 2×96-well pre-coated strip plates (6 strips/48 wells per neurotrophin target), protein standards, detection reagents, wash and sample buffers, substrate buffer and detailed protocols.
Caution: For research use only. Not for diagnostic and clinical purposes.
Customers Who Bought This Also Bought