To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Gene ID:333932
- Protein/peptide type:Recombinant
- Source:Wheat germ
- Species:Human
- Conjugation:Unconjugated
- Size:2 μg
- Storage conditions:Store at -80C. Avoid freeze-thaw cycles.
- Protein synonyms:H3F2|H3FM|H3a|Histone H3/m|H3/n|histone 2|histone cluster 2|HIST2H3D|Histone H3/o|histone H3.2|HIST2H3C|H3/o
- Protein/peptide name:HIST2H3A
- Purity:>80% by SDS-PAGE and Coomassie blue staining
- Cat. no.:10521-122
- Supplier No.:H00333932-P01
Specifications
About this item
The Recombinant Human HIST2H3A Protein from Novus Biologicals is derived from Wheat germ. The Recombinant Human HIST2H3A Protein has been validated for the following applications: Western Blot, ELISA, SDS-Page, Protein Array, Immunoaffinity Purification.