Radiation Can Set
Catalog #: 470304-032
Supplier: United Scientific Supplies
Specifications
- Description:Radiation Can Set
- Cat. no.:470304-032
Specifications
About this item
High Quality for the Classroom.
- Includes activity guide
Two identical sized screw cap metal cans (one smooth metal and other painted dull black) to demonstrate the difference in cooling rates of hot water inside the cans due to different surfaces.
The difference in the cooling rates is attributed to the thermal radiation emitted by the two cans.
Customers Who Bought This Also Bought