Specifications
- Description:SafeCup™ III MultiDrug 14 Panel
- Size:25 Cups/Box
- Cat. no.:76266-174
Specifications
About this item
SafeCup™ lll Round Cups are an all-in-one (collection/testing/transport) qualitative test for the detection of drugs of abuse in urine.
- Tests adhere to SAMSHA cutoff levels
- Leak-proof cap, temperature strip, & security seal included
All-in-One Cup tests for AMP, BAR, BZO, BUP, COC, MTD, MDMA, MET, OPI, OXY, PCP, TCA, THC, and PPX. CLIA Waived. Includes 25 collection cups, security seals, gloves, and instructions.