Quantum Levitation Classroom Kit
Catalog #: 470306-210
Supplier: QUANTUM LEVITATION LTD
Specifications
- Description:Quantum Levitation Classroom Kit
- Cat. no.:470306-210
Specifications
About this item
The Classroom Kit will allows students to experiment with modern physics and learn about one of the most intriguing and exciting phenomenon of modern times – Superconductivity.
- Easy to use
- Highly durable
The kit contains a set of easy to conduct student experiments suitable for both high-school and undergraduate levels. The experiments and data analysis will enrich your student’s understanding and give them a taste of the work of true scientists.
Demonstrations and Student Activities:
Quantum Levitation.
The Meissner Effect.
Quantum Locking.
The Maglev Track.
Quantitative Experiments:
Measuring the Levitation Force.
Measuring the Critical Temperature Using Magnetic Levitation.
Superconductivity as a Mean to Investigate Classical Mechanics.
Ordering information: The Classroom Kit comes with Round maglev track Ø 40 cm (Ø15.7”)); Wooden stands; 3 handheld magnetic device; 1 medium Quantum Levitators; 1 small Quantum Levitators; 3 small Quantum Ed Levitators (thermometer included); Printed copy of “Quantum Levitation Booklet for Teachers“; and User manual.
Customers Who Bought This Also Bought