Quantum Levitation Maglev Kit
Catalog #: 470306-204
Supplier: QUANTUM LEVITATION LTD
Specifications
- Description:Quantum Levitation Maglev Kit
- Cat. no.:470306-204
Specifications
About this item
The maglev kit is perfect for large audience demonstrations at science museums, science fairs and student classes.
- Easy to use
- Highly durable
The circular frictionless motion never seizes to amaze.
When used with the new resistance thermometer equipped Ed levitator, the handheld magnetic device serves as a unique experimentation platform where quantum phenomena such as the Meissner effect and flux pinning can be investigated.
Delivery: The kit is supplied with large and medium Quantum Levitators which can be levitated simultaneously in a double levitation configuration. Both suspension below the track and levitation above it can be demonstrated. A handheld magnetic device (Starter Kit) is also included for a complete experience.
Customers Who Bought This Also Bought